DNA methylation kit, Express (deep well)
Catalog # ENZOENZ450020002
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Express DNA methylation kit (deep well), 2×96 wells
- Sekvence:02
Specifications
About this item
The Express DNA Methylation Kit (deep well) features high-throughput (96-well) bisulfite treatment and conversion of DNA for methylation analysis.
- Ready to use conversion reagent is added directly to DNA
- Easy to follow instructions
- High yield
Desulphonation and clean-up of the converted DNA is performed using a 96-well spin-plate. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Delivery: Kit contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, L-Desulphonation reagent, Methylation elution buffer, Express binding plate (deep well), Express conversion plates, collection plates and elution plates.