Clostridium difficile toxin A
02, Clostridium difficile toxin A, 50 µg, 50 µG
Catalog # ENZOBMLG1400050
Supplier: Enzo Life Sciences
<i>Clostridium difficile</i> toxin A
Catalog # ENZOBMLG1400050
Supplier: Enzo Life Sciences
CAS Number:
Jump to:
Product Details & Documents
Clostridium difficile toxin A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies and LINE assay.
- Minimal lot to lot variation
- Single lot bulk quantities available
- Lyophilised for ease of handling and long term stability
Specifications
Specifications
- Bal.:50 µG
- Popis:Clostridium difficile toxin A, 50 µg
- Sekvence:02