Clostridium difficile toxoid A
Catalog # ENZOBMLG1450050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Bal.:50 µG
- Popis:Clostridium difficile toxoid A, 50 µg
- Sekvence:02
Specifications
About this item
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
- Minimal lot-to-lot variation
- Inactivated by formaldehyde
- Lyophilised for ease of handling and long term stability