Clostridium difficile toxin B
Catalog # ENZOBMLG1500050
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Bal.:50 µG
- Popis:Clostridium difficile toxin B, 50 µg
- Sekvence:02
Specifications
About this item
Clostridium difficile toxin B from Clostridium difficile, strain VPI10463 is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies and LINE assay.
- High potency retained following inactivation
- Minimal lot-to-lot variation
- Lyophilised for ease of handling and long term stability