- Druh protilátky:Primární
- Název antigenu:Tyrosinase
- Symbol antigenu:TYR
- Klonovalita:Monoclonal
- Klon:Clone T311 OCA1/812
- Spojení:Unconjugated
- Hostitel:Mouse
- Imunochemie:Yes
- ImunoFluorescence:Yes
- Izotyp:IgG
- Reaktivita:Human
- Identifikační číslo genu:503555
- Synonyma antigenu:Tumor rejection antigen AB|ATN|CMM8|SHEP3|Oculocutaneous albinism IA|Monophenol monooxygenase|OCA1A|OCA1|TYR|LB24-AB|SK29-AB
- Skladovací pufr:PBS/0,05% azide
- Koncentrace:1 mg/mL
- Čištění:BSA-free
- Rozměry protilátky:50 μL
- Bal.:50 µl
Recognizes a cluster of proteins between 70-80 kDa, identified as tyrosinase. Occasionally a minor band at 55 kDa is also detected. This MAb shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. Staining of melanomas with this MAb shows tyrosinase in melanotic as well as amelanotic variants. This MAb is a useful marker for melanocytes and melanomas.
Type: Primary
Antigen: TYR
Clonality: Monoclonal
Clone: T311 OCA1/812
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human