Order Entry
Czech Republic
ContactUsLinkComponent
Anti-VWF Mouse Monoclonal Antibody (Purified, BSA-free) [clone: VWF635]
Anti-VWF Mouse Monoclonal Antibody (Purified, BSA-free) [clone: VWF635]
Catalog # BTIUBNUM0635-50
Supplier:  Biotium
Anti-VWF Mouse Monoclonal Antibody (Purified, BSA-free) [clone: VWF635]
Catalog # BTIUBNUM0635-50
Supplier:  Biotium

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    von Willebrand Factor
  • Symbol antigenu:
    VWF
  • Klonovalita:
    Monoclonal
  • Klon:
    Clone VWF635
  • Spojení:
    Unconjugated
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • Izotyp:
    IgG1 kappa
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Identifikační číslo genu:
    440848
  • Synonyma antigenu:
    F8VWF|von Willebrand Disease (vWD)|Factor VIII Related Antigen|Coagulation Factor VIII|von Willebrand Antigen 2
  • Skladovací pufr:
    PBS/0,05% azide
  • Koncentrace:
    1 mg/mL
  • Čištění:
    BSA-free
  • Rozměry protilátky:
    50 μL
  • Bal.:
    50 µl

Specifications

About this item

von Willebrand Factor (vWF) is a multimeric glycoprotein that is found in endothelial cells, plasma and platelets. It acts as a carrier protein for Factor VIII and promotes platelet adhesion and aggregation. vWF undergoes a variety of posttranslational modifications that influence the affinity and availability for Factor VIII, including cleavage of the propeptide and formation of N-terminal disulfide bonds. This antibody helps to establish the endothelial nature of some lesions of disputed histogenesis, e.g. Kaposi's sarcoma and cardiac myxoma. It is widely used for differentiating vascular lesions from those of other tissue differentiation within a panel of other vascular markers although not all tumors of endothelial differentiation contain this antigen.

Type: Primary
Antigen: VWF
Clonality: Monoclonal
Clone: VWF635
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human