Visual colour kits, magnesium, MQuant®, Supelco®
1.11131.0001
: Merck
:
- Chemická metoda:Xylidyl blue
- Bal.:50 Tests
- Popis:Magnesium test
- Sekvence:02
- M-Clarity quality level:MQ100
- Typ:MTC
- Rozsah:100-200-300-500-1000-1500 mg/l Mg
Quick tests, visual colour comparison.
- Results in approximately 2 minutes
- Easy to use
- Application-oriented measuring ranges
- Portable
MQuant® titrimetric and colorimetric methods:
This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.
: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.