Anti-BrdU Mouse Monoclonal Antibody [clone: 2B1]
ENZOADIMSA200E
- Druh protilátky:Primární
- Název antigenu:Bromodeoxyuridine
- Symbol antigenu:BrdU
- Klonovalita:Monoclonal
- Klon:2B1
- Spojení:Unconjugated
- Průtoková cytometrie:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- Izotyp:IgG1
- Reaktivita:Multiple
- Sekvence:02
- Formulář:Liquid
- Skladovací pufr:PBS, pH 7,4, containing 10 mg/ml sucrose and 1,0 mg/ml sodium azide
- Skladovací teplota:−20 °C
- Transportní teplota:Shipped on blue ice
- Imunogen:Iododeoxyuridine-Ovalbumin
- Čištění:Purified by Protein A affinity chromatography
- Bal.:100 µG
The thymidine analogue BrdU (5-Bromo-2-Deoxyuridine), a derivative of uridine, can substitute for thymidine during DNA synthesis.
- Dilution range: Flow Cytometry (10 µg/ml), Immunohistochemistry (10 µg/ml)
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: 2B1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species independent