Labels for BMP™ 51/53 printer, type B-595
Katalogové číslo BRDY170949
Dodavatel: Brady
Specifikace
- Type:B-595
- Barva:Černá/bílá
- Rozměry:6,35 mm×7,62 m
- Materiál:Vinyl
- Sekvence:40
- Model:M4C-250-595-WT-BK
- Balené:1 label/box
- Bal.:1
- Adhezní:Permanentní akryl
Specifikace
O tomto produktu
These are made from durable, low-shrink vinyl with a very aggressive adhesive. The labels adhere to irregular, curved, rough and highly textured surfaces (painted cinder blocks, uneven wood, textured plastics, paper jacketed pipes, powder coated surfaces). Also adhere to difficult low energy surface items (PVC piping, blow-moulded equipment cases, ABS plastics, recycled plastics).