Digital wood moisture analyser
Catalog # SIMELI7000
Supplier: Industrial Physics
Specifications
- Rozlišení:0,1 %
- Popis:Digital wood moisture analyser
- Rozsah vlhkosti:7 – 40 %
- Š×H×V:150×80×30 mm
- Bal.:1
Specifications
About this item
Non destructive instrument for measuring moisture levels in wood.
- Calibration lines for 25 different types of wood
- High degree of accuracy
- Automatic power off
The selected wood and the measuring results are immediately shown. The instrument can also detect the moisture content of concrete, anhydrite, estrich, polyester.
Delivery: Supplied in a pouch.