No Cal stain lifter solution, Orion™
No Cal stain lifter solution, Orion™, 60 ml bottle
Katalogové číslo ORIO510015
Dodavatel: Thermo Orion
Přejít na:
Product Details & Documents
This cleaning solution restores proper electrode performance and prolongs the useful life of the electrode.
Specifikace
- Popis:No Cal stain lifter solution, Orion™, 60 ml bottle