Anti-ACADVL Rabbit Polyclonal Antibody
Catalog # BSBTPB10023
Supplier: Boster Bio
Specifications
- Druh protilátky:Primární
- Název antigenu:Acyl-Coa Dehydrogenase, Very Long Chain
- Symbol antigenu:ACADVL
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat
- Western blot:Yes
- Křížová adsorbce:No
- Identifikační číslo genu:37
- Synonyma antigenu:LCACD|VLCAD|ACAD6
- Rozměry protilátky:100 μg
- Bal.:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Very long-chain specific acyl-CoA dehydrogenase, mitochondrial(ACADVL) detection. Tested with WB in Human;Rat.
Type: Primary
Antigen: ACADVL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat