Labels for minimark printers, type B-7563, MetaLabel
Catalog # BRDY710081
Supplier: Brady
Specifications
- Type:B-7563
- Barva:Stříbrná
- Popis:Printer labels
- Rozměry:25×12 mm
- Materiál:Polyester
- Model:MNK-BPT-5-7563
- Balené:2865 labels/roll
- Bal.:1 Roll
- Adhezní:Permanentní akryl
Specifications
About this item
Metallic looking labels for identifying asset and rating plates.