ContactUsLinkComponent
Modular assemblies for laboratory fume cupboards
Modular assemblies for laboratory fume cupboards
Catalog # KOTT405-00028
Supplier:  KOETTERMANN
undefined
Modular assemblies for laboratory fume cupboards
Catalog # KOTT405-00028
Supplier:  KOETTERMANN
Specifications
  • Popis:
    Variable assembly for high-performance fume cupboard with ceramic lining – HFC (Cer) and HFCr (Cer), standard, low structure, knocked out, 2100 mm
  • Sekvence:
    01
  • Š×H×V:
    2100×2500 mm
  • Bal.:
    1
Specifications
About this item
Modular fume cupboards are assembled from many different modules. The characteristic assembly for each type is called a 'variable assembly'.
  • One variable assembly is required for each modular fume cupboard
Fume cupboards are basically available with two different shipping options, which are part of the variable assemblies:
Knocked down: The front, sash, casing and support frame are shipped as assemblies and must be put together on site. As a rule, knocked-down fume cupboards of all types can be carried into laboratories even if the access is narrow.
Partially assembled: The front and sash are assembled in the factory and shipped as a complete front. As a rule, partial assembly is advantageous because it reduces installation time on site. In many cases fume cupboard fronts up to 1500 mm wide can be carried in on site.
Please contact Avantor for further details.