Orders LinkContactUsLinkComponent
Buckets, Rectangular, PP
Buckets, Rectangular, PP
Certifikáty
Buckets, Rectangular, PP
5 Dostupných variant níže
Product Details & Documents
Standard for Clino® system trolleys, rectangular bucket with handle. Also suitable for the pre-preparation of textiles.
  • With scaling from 1 to 5 L
  • Coloured to facilitate use as part of a colour coded system
  • Good chemical resistance
  • Good temperature resistance
Product Family Options
Informace o produktu
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Cover for 6 l buckets, transparent
    Cover for 6 l buckets, transparent
    PPSA2000610
    01
    Přihlásit se pro dostupnost
    1 * 1 KS - 607,00 Kč