Test kit, AF129CYS, balanced water including cyanuric acid, Comparator 2000+
Catalog # LOVI411296
Supplier: Lovibond
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Balanced water cyanuric acid test kit
- Typ:AF129CYS
- Rozsah:0 - 4 mg/l (Cl2); 6,8 - 8,4 pH; 10 - 500 mg/l (total alkalinity); 10 - 500 mg/l CaCO3 (calcium hardness); 20 - 200 mg/l (cyanuric acid)
Specifications
About this item
Comparator 2000+ is a versatile modular system for testing water.This kit measures chlorine, alkalinity, pH calcium hardness and cyanuric acid.
- Stable and accurate
- Robust, compact and portable
- Prism enabling colour matching
- Diffuser plate is used to measure the turbidity
- Longer path length cells be used for lower concentration analysis
Delivery: Test kit is supplied in a robust carrying case containing Comparator 2000+, pH comparator disc, chlorine comparator disc, 4× comparator vials, 10 ml, 13 mm path length, shaker tube, cyanuric acid tube, tablet reagents for each parmeter, crushing/stirring rod, vial cleaning brush, 100 ml graduated cylinder, instruction sheet, langelier index/balanced water and calculation table.