Bouffant caps, Tyvek® IsoClean®, model IC 729 S
Dodavatel: DuPont
Certifikáty
O tomto produktu
These caps are made of Tyvek® which is a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
- Ultra-low linting fabric
- Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
- Elastic headband
- Garments are available non sterile or gamma sterilised to SAL of 10⁻⁶
- Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio (also available non clean-processed)
Ideal for use in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments.
Certifications: (EU) 2016/425; Cat III, Type PB [6-B]; EN 14126
Packaging: 250 per box, bulk packaged in double transparent poly liners.
Specifikace
- Catalog No:
- 113-8646
- DUPOIC729SWH00
- Aplikace oblečení:
- Cleanroom
- Cleanroom
Specifikace
Související informace
Cleanroom_Whitepaper_Advantages of IsoClean_EN_Final DUPONT™ TYVEK® ISOCLEAN®Sizing Chart Accessories
Application Notes
Controlled_Environments_Catalogue_2016_EN DUPONT™ TYVEK® ISOCLEAN® DUPONT™ TYVEK® ISOCLEAN® FAQ Poster Non-Sterile Cleanroom Gowning Procedure
Product Literature
DUPOIC729SWHMS_TDS_EN
Datasheet
Frequently Bought Together