Capillary columns, HP-fast residual solvent
HEWL19095V-420LTM
: Agilent
- Formát kolony:Capillary
- Tloušťka filmu:1,00 µm
- Ø vnitř.:0,53 mm
- Délka:30 m
- Sekvence:03
- Fáze:HP-Fast Residual Solvent
- Bal.:1
Bonded and cross-linked GC columns.
- Equivalent to USP Phase G43
- Thinner film reduces run time by 2,5 times and increases MDL by 2 times compared to standard film thickness