Digital tensiometer, TE 3
461-0750
: LAUDA
- Model:TE 3
- Rozsah teplot:5…80 °C (+0,1) with PTT (optional)
- Popis:Digital tensiometer
- Pohyb stolku:Motor driven from 0,1 to 1 mm/second in ten steps
- Pnutí povrchu/rozhraní:Du Nouy ring: 0 - 300 mN/m ; Plate: 0 - 999 mN/m ; Resolution: 0,01 mN/m
- Rozsah vážení:0 - 5000 mg
- Š×H×V:245×205×335 mm
- Rozsah hustoty:0 - 2000 g/L
- Napětí:Ring: 0 - 300 mN/m ; Platte: 0 - 999 mN/m ; Platte: 0,01 mN/m
- Hmotnost:5 kg
- Bal.:1
Using a Du Noüy ring or Wilhelmy plate, the TE 3 can be combined with a Peltier thermostatting stirring unit (PTT).
This digital tensiometer allows for surface, interfacial tension, and density.
: DIN 53914 and 53993, ASTM D971, ISO 304 and 4311.
: Supplied with two-branch Du Noüy ring, weight calibration (500 mg), glass plunger (for density), pliers, 10× glass beakers (Ø = 6 cm) and case.