Orders LinkContactUsLinkComponent
Precision scale magnifiers
03, Magnifier with battery LED handle (Ø = 35 mm), 38,0 dioptre, 10×
  ESCH155074
Precision scale magnifiers
  ESCH155074
Precision scale magnifiers
Precision scale magnifiers
03, Magnifier with battery LED handle (Ø = 35 mm), 38,0 dioptre, 10×
4 370,00 Kč
1 * 1 KS
Precision magnifiers with brightness control and removable measuring scale available with battery or mains handle with brightness control.
Aspheric PXM® lightweight lens with Cera-tec® hard coating (both sides).
  • Zvětšení:
    10×
  • Popis:
    Magnifier with battery LED handle (Ø = 35 mm), 38,0 dioptre
  • Sekvence:
    03