Hand brushes and brooms, PP
PPSA3590300
- Barva štětin:Bílá
- Materiál štětin:PBT
- Popis:Hand broom, 310×40×90 mm
- Bal.:1
- Handle colour:Bílá
White hand broom with run direction for hygienically sensitive areas to clean difficult-to-access areas in cleanrooms. Material: PP (body), PBT (trimming).
- Stock height 45 mm
- Unslit fiber ends
- Hard bristle material