Orders LinkContactUsLinkComponent
Digital timer
Digital timer
Electronic timer, Bílá, 68×20×53 mm
Catalog # AMARE920630
Supplier:   Amarell
Digital timer
Catalog # AMARE920630
Supplier:  Amarell
Digital timer
Electronic timer, Bílá, 68×20×53 mm
Price
631,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Count down/count up timer with clock and stopwatch.
  • Timing capacity: 24 hours
  • Loud alarm signal
  • Memory function
  • With magnet, standing and holding device
Delivery information: Supplied with round cell LR44/A76.
Specifications
  • Barva:
    Bílá
  • Popis:
    Electronic timer
  • Š×H×V:
    68×20×53 mm