ContactUsLinkComponent
Syringe filters, Celtron, Whatman™
Syringe filters, Celtron, Whatman™
Katalogové číslo 514-1138
Syringe filters, Celtron, Whatman™
Katalogové číslo 514-1138
Specifikace
  • Ø:
    30 mm
  • Velikost pórů:
    0,45 µm
  • Type:
    GF92-TG100
  • Sekvence:
    04
  • Bal.:
    100
Specifikace
O tomto produktu
Celtron ready-to-use filter units have an integral glass fibre prefilter to retain coarse particles and a membrane filter for filtering the fine contaminants.
  • Regenerated cellulose membranes have an excellent chemical resistance to organic solvents
  • Cellulose acetate membranes have an extremely low binding capacity for proteins