Crotonoyl chloride cis- and trans-mixture for synthesis, Sigma-Aldrich®
Catalog # 8.20351.0025
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:25 ml
- Sekvence:2315
Specifications
About this item
:
:104.53 g/mol
ADR:8 (3),II
:M-Clarity™ quality level = MQ200
EINECS:234-010-3
UN:2920
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:10487-71-5
:625-35-4
Formula:C₄H₅ClO
MDL Number:MFCD00000734