Inline check valves, quick-stop luer, Upchurch Scientific®
Katalogové číslo 554-3155
Dodavatel: UPCHURCH SCIENTIFIC
Specifikace
- Popis:P-696 Quick-Stop Luer check valve assembly, 127 µl swept volume
- Materiál:PEEK
- Bal.:1
Specifikace
O tomto produktu
The Quick-Stop Luer check valve assembly is specifically designed to provide inline Luer connect/disconnect convenience while avoiding spills. It is made from biocompatible PEEK with gold-plated stainless steel spring.
- Remains open when engaged
- Designed to provide inline Luer connect/disconnect convenience without the mess and hazard of spills