Foam hand sanitiser dispenser, InstantFOAM TouchFREE
Catalog # EVONIFSTF2MD-UK
Supplier: Deb-STOKO
Specifications
- Kapacita:1000 ml
- Barva:Bílá
- Popis:Foam hand sanitiser dispenser, InstantFOAM TouchFREE, plastic
- Š×H×V:170×100×277 mm
- Bal.:1
Specifications
About this item
Energy-efficient touch-free dispensing system for use with foam hand cleansers.
- Minimises any risk of cross-contamination
- 4 D-cell batteries provide up to five years of operation
- Infrared sensing technology to prevent misfiring
- BioCote® protection antimicrobial agent inhibits the growth of bacteria and moulds
- Translucent covers provide visibility of the cartridge without opening the dispenser
- Dispenser can be locked and opened with the special key provided, can also be easily converted to a keyless, non locking operation
- For wall-mounting
Suitable for use in public facility washroom, offices, medical rooms, laboratories, repair shops, food service and catering areas with InstantFOAM® cartridges.