Syringe priming kit
Katalogové číslo HAMIPRMKIT
Supplier: HAMILTON BONADUZ
Specifications
- Popis:Priming kit
- Bal.:1 KIT
Specifications
About this item
The priming kit provides the necessary tools to ensure that the syringe and needle connection is completely primed. The glass capillary tubing traps a large volume of liquid that can only be overcome by using the priming kit. The kit is compatible with 600, 700, 1700, removable needle syringes and 7000 series syringes.
Delivery: Kit includes one 250 µl syringe, one 6 pack of 30 gauge needles and one pack of septa.