Orders LinkContactUsLinkComponent
Fixation buffer for microscopy and flow cytometry
06, Fixation buffer, ready to use, 100 mL
Catalog # 22015.
Supplier:   Biotium
Fixation buffer for microscopy and flow cytometry
Catalog # 22015.
Supplier:  Biotium
CAS Number:  
Fixation buffer for microscopy and flow cytometry
Fixation buffer for microscopy and flow cytometry
06, Fixation buffer, ready to use, 100 mL
CAS Number:  
Price
4 930,00 Kč
Log in for availability
1 * 100 mL
Product Details & Documents
Ready-to-use formaldehyde-based fixation buffer for immunofluorescence staining for microscopy or flow cytometry.
Specifications
Specifications
  • Bal.:
    100 mL
  • Popis:
    Fixation buffer, ready to use
  • Sekvence:
    06
  • Cat. No.:
    22015.
Specifications