Syringes, 3-piece, HENKE-JECT®
Catalog # 613-2793
Supplier: HENKE SASS WOLF
Specifications
- Kapacita:1 ml
- Popis:Syringes, Luer lock
- Bal.:100
Specifications
About this item
These syringes are commonly used for embryo transfer and oocyte handling.
- Rubber piston syringes
- Double seal for increased safety
- Pyrogen-free, PVC-free, non-toxic
Syringes have smooth flow, tight and high transparency barrel and the lubricated synthetic rubber piston allows fine dosing.