Natronové vápno, granular, pure indicator, with indicator
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8006-28-8
:
MDL Number:MFCD00134094
ADR:8,III
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige to light grey Granules
- Loss on drying:≤20 % (10 g, 200°C, 2 h)
- CO2 absorption capacity:≥19.0 %
Specification Test Results
Frequently Bought Together