Microcentrifuge tubes
Katalogové číslo MLBP514
Supplier: Thermo Fisher Scientific
Specifications
- Type:with thread, without screw cap
- Verze:graduated, conical bottom, non sterile
- Kapacita:1,5 ml
- Barva:Bezbarvá
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Bal.:500
- Max. RCF:20000×g
Specifications
About this item
PP, translucent.
- Frosted writing area on tube and cap