Orders LinkContactUsLinkComponent
pH electrodes, Eutech
pH electrodes, Eutech
 :   EUTECH
pH electrodes, Eutech
 :  EUTECH
pH electrodes, Eutech
pH electrodes, Eutech
pH electrodes for use with all pH meters with BNC input connector.
Product Family Options
  • Sekvence
    Popis
    pH
    Tepl.
    Materiál
    Diafragma
    Ø×D
    Připojení
    Určeno pro
  • 01
    pH electrode, single junction, sealed
    1 - 13
    0...+80 °C
    Epoxidová pryskyřice
    Porous HDPE pin
    12×90 mm
    BNC, 1 m cable
    General purposes
    1 * 1 KS
    1 * 1 KS - 2 480,00 Kč
    2 480,00 Kč
    1 * 1 KS
       2 480,00 Kč
  • 01
    pH electrode, double junction, sealed
    1 - 13
    0...+80 °C
    Epoxidová pryskyřice
    Porous HDPE pin
    12×90 mm
    BNC, 1 m cable
    General purposes
    1 * 1 KS
    1 * 1 KS - 3 330,00 Kč
    3 330,00 Kč
    1 * 1 KS
       3 330,00 Kč
  • 03
    pH kombinovaná elektroda s dvojitým můstkem, těsnicí, zabudovaná ATC
    1 - 13
    0...+80 °C
    Epoxidová pryskyřice
    Porézní HDPE
    12×115 mm
    BNC
    , 1m kabel
    Obecné použití
    1 * 1 KS
    1 * 1 KS - 4 620,00 Kč
    4 620,00 Kč
    1 * 1 KS
       4 620,00 Kč
  • 04
    pH combination electrode, single junction, refillable, with protective sensor guard
    0 - 13
    0...+100 °C
    Sklo
    Annular ceramic
    12×130 mm
    BNC, 1 m cable
    General purposes (samples with low temperature measurements)
    1 * 1 KS
    1 * 1 KS - 3 610,00 Kč
    3 610,00 Kč
    1 * 1 KS
       3 610,00 Kč
  • 05
    pH combination electrode, double junction, refillable, clog-free
    0 - 13
    0...+100 °C
    Sklo
    Annular ceramic
    12×110 mm
    BNC, 1 m cable
    Samples containing TRIS buffers, sulfides and more
    1 * 1 KS
    1 * 1 KS - 7 430,00 Kč
    7 430,00 Kč
    1 * 1 KS
       7 430,00 Kč
  • 06
    pH electrode, rugged, single junction, sealed
    0 - 14
    5...+110 °C
    Sklo
    Annular ceramic
    12×110 mm
    BNC, 1 m cable
    Strong alkaline solutions, photographic chemicals
    1 * 1 KS
    1 * 1 KS - 7 240,00 Kč
    7 240,00 Kč
    1 * 1 KS
       7 240,00 Kč
  • 07
    pH combination electrode, single junction, sealed
    0 - 13
    0...+100 °C
    Sklo
    Annular ceramic
    8×55 mm
    BNC, 1 m cable
    Solid / semi-solid samples
    1 * 1 KS
    1 * 1 KS - 7 240,00 Kč
    7 240,00 Kč
    1 * 1 KS
       7 240,00 Kč
  • 08
    pH electrode, single junction, refillable (with solution ECRE001)
    0 - 13
    0...+100 °C
    Sklo
    Annular ceramic
    8×55 mm
    BNC, 1 m cable
    High viscosity samples where frequent cleaning of reference is required
    1 * 1 KS
    1 * 1 KS - 6 030,00 Kč
    6 030,00 Kč
    1 * 1 KS
       6 030,00 Kč