Orders LinkContactUsLinkComponent
First aid kit, medium
First aid kit, medium
First aid kit, medium, durable EVA case (green/yellow), with handle, 231×78×190 mm
Katalogové číslo 118-0394
Dodavatel:   ORKLA CARE
First aid kit, medium
Katalogové číslo 118-0394
Dodavatel:  ORKLA CARE
First aid kit, medium
First aid kit, medium
First aid kit, medium, durable EVA case (green/yellow), with handle, 231×78×190 mm
Přejít na:
Price
2 100,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
This portable first aid kit is designed for personal use. It contains first aid products to be used in emergency situations (bleeding, breathing, burns, sprains, circulatory shock). Ideal for people who are often on the go and as a complement to a wall-mounted first aid station.
  • Strong and durable case made from a rubber-like material (EVA) with a mat finish
  • Handle with adjustable Velcro® strap makes it easy to carry the kit around
  • Products packed in transparent plastic pockets: Easy to find the product that is needed
  • All products protected against dust, dirt and moisture
  • Products come with clear, self-explanatory instructions
  • Enough space for additional products
Contents:
1× Cederroth 4-in-1 Bloodstoppers
1× Cederroth 4-in-1 mini Bloodstoppers
10× Salvequick plasters
6× Salvequick wound cleansers
4× Salvequick Maxi cover
1× Breathing mask
2× Safety hand cleanser
2× Pairs of gloves
1× Rescue blanket
1× Elastic bandages
1× Cederroth burn gel dressing
1× Pair of scissors
1× First aid instructions
Shelf life: 5 years from production date for each component
Specifikace
  • Popis:
    First aid kit, medium, durable EVA case (green/yellow), with handle
  • Rozměry:
    231×78×190 mm