Caps for cell culture flasks, EasYFlasks™, Nunclon™Δ
Catalog # 734-2144
Supplier: Thermo Fisher Scientific
Specifications
- Popis:Vent/close caps for 175 cm² flasks
- Bal.:100
Specifications
About this item
Ideal for cell culture flasks. Offered in two different types vent/close and filter.
- Sterile
- HDPE closure