ContactUsLinkComponent
Controlled Protein-Protein Crosslinking Kit, Pierce™
Controlled Protein-Protein Crosslinking Kit, Pierce™
Catalog # PIER23456
CAS Number:  
undefined
Controlled Protein-Protein Crosslinking Kit, Pierce™
Catalog # PIER23456
CAS Number:  
Specifications
  • Bal.:
    1 KIT
  • Popis:
    Controlled Protein-Protein Crosslinking Kit, sufficient for one conjugation with two purified proteins (up to 5 mg each)
Specifications
About this item
The Pierce™ Controlled Protein-Protein Crosslinking Kit contains essential crosslinkers, modification reagents and buffers for conjugating antibodies, enzymes and other purified proteins needed as probes.
  • Reliable non cleavable heterobifunctional crosslinking agent offers proven chemistry, highly stable intermediates and efficient formation of the target conjugate
  • Includes disulfide (S-S) reductants and thiolation reagents
  • Ellman’s Reagent provides the option to monitor reaction sufficiently
Delivery: The kit includes Sulfo-SMCC crosslinking agent (2 mg); 10X activation/conjugation buffer (20 ml); BupH phosphate buffered saline (2 packs); 2-Mercaptoethylamine-HCI (6 mg); immobilised reductant (0,2 ml); SATA (2 mg); Dimethylformamide (DMF, 1 ml); Hydroxylamine-HCI (5 mg); dextran desalting columns (2×10 ml); column extender (2 each); cysteine-HCI (20 mg); and Ellman’s reagent (2 mg).