PCR plate, 96-well, automation compatible, Axygen®
731-0678
: Corning
- Poč. jamek:96
- Lem:Semi-skirted
- Well colour:Průhledná
- Popis:PCR plate, conical well bottom
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:I-RECs, ISO 14001 - Reynosa
- Bal.:10
- Objem jamky:0,2 ml
This automation-compatible, polypropylene PCR microplate features a rigid frame with a small split that sustains heat transferring during and post thermal cycling without distorting its shape. The plate also features raised rims around the wells to provide tight sealing to minimise heat evaporation.
- Remains flat during and post thermal cycling
- Raised well rims help reduce heat evaporation
- Clear wells, visible grid for easy sample loading and tracking
- Ultra-thin and consistent wall thickness for easy and consistent heat transfer
- Single notch corner
- SBS footprint
- Free from RNase and DNase, non pyrogenic
Compatible with most major thermal cyclers, sequencers, and automation platforms, the plate provides optimal performance for downstream applications, including high throughput screening.