Test kits, monochloramine, Spectroquant®, Supelco®
1.01632.0001
: Merck
:
- Chemická metoda:Indophenol blue
- Bal.:150 Tests
- Popis:Monochloramine test
- M-Clarity quality level:MQ100
- Pro jednotku:Prove, Nova 60, Multy, Move
- Rozsah:0,050 - 10,00 mg/l Cl₂, 0,036 - 7,25 NH₂Cl, 0,010 - 1,96 NH₂Cl - N
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.