Test kits, total hardness cell test, Spectroquant®, Supelco®
Catalog # 1.00961.0001
Supplier: Merck
CAS Number:
Specifications
- Chemická metoda:Phthalein complexone
- Bal.:25 Tests
- Popis:Total hardness cell test
- M-Clarity quality level:MQ100
- Pro jednotku:Prove, Nova 30/60, Multy, Move
- Rozsah:5 - 215 Ca, 0,7 - 30,1 °d, 0,9 - 37,6 °e, 1,2 - 53,7 °f, 7 - 300 mg/l CaO, 12 - 537 CaCO₃
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.