ContactUsLinkComponent
Phosphoprotein enrichment kit, Pierce™
Phosphoprotein enrichment kit, Pierce™
Catalog # PIER90003
CAS Number:  
undefined
Phosphoprotein enrichment kit, Pierce™
Catalog # PIER90003
CAS Number:  
Specifications
  • Bal.:
    10
  • Popis:
    Phosphoprotein enrichment kit, 10×1 ml columns
Specifications
About this item
Pierce™ Phosphoprotein Enrichment Kit is designed for the efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues.
  • Low non specific protein contamination from complex biological samples
  • Easy spin format enables enrichment of phosphorylated proteins in less than 2 hours
  • High yields
Pierce™ Phosphoprotein Enrichment Kit uses a convenient spin column format and is compatible with downstream applications including Western blotting, mass spectronomy, 2D-PAGE, and protein arrays.