Streptavidin poly-HRP conjugate, Pierce™
Catalog # PIER21140
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Bal.:0,5 mL
- Popis:Streptavidin Poly-HRP conjugate
Specifications
About this item
Pierce™ Streptavidin Poly-HRP conjugate is biotin-binding protein conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for IHC and other methods.
- Detect low-abundant targets (low picogram-femtogram range) with high signal-to-noise ratios
- Compatible with chromogenic, fluorogenic and chemiluminescent substrates, also with ELISA, Western blotting and immunohistochemistry (IHC)
- Can be directly substituted into immunoassays and other detection assays
- Requires less conjugate per assay and standard HRP conjugates