ContactUsLinkComponent
Cylinders for urinometers
Cylinders for urinometers
  612-2764
undefined
Cylinders for urinometers
  612-2764
  • Ø:
    28 mm
  • Výška:
    175 mm
  • Sekvence:
    901
  • Bal.:
    1
Jar, made of glass.
With base and spout for urinometers.