ContactUsLinkComponent
Spin filter plates, 96 well, for Ni-NTA proteins, HisPur™
  PIER88230
undefined
Spin filter plates, 96 well, for Ni-NTA proteins, HisPur™
  PIER88230
  • Popis:
    HisPur™ Ni-NTA Spin Plates, 2×96-well
  • Bal.:
    1 KIT
The HisPur™ Ni-NTA spin plates contain a high-capacity, high-performance nickel-IMAC resin for routine affinity purification of His-tagged fusion proteins.
  • High capacity binding up to 60 mg of 6xHis-tagged protein per ml of resin
  • Purify proteins using native or denaturing conditions
  • Resin can be reused at least five times without reduction in performance
  • Purifies high levels of overexpressed His-tagged fusion proteins from bacterial lysates
These spin plates are ideal for simultaneous processing of multiple small (1 mg/well binding capacity) samples. The specially prepared support consists of beaded agarose derivatised with the nitrilotriacetic acid (NTA) chelation moiety and loaded with divalent nickel ions (Ni²⁺). The immobilised metal affinity chromatography (IMAC) resin provides exceptional binding capacity and performance for recombinant His-tagged protein purification.
 : Includes two filter plates with each well containing 50 µl of resin bed (200 µl of 25% resin slurry), and six wash/collection plates.