ContactUsLinkComponent
Protein quantitation reagent, TMTzero™ label reagent
Protein quantitation reagent, TMTzero™ label reagent
Catalog # PIER90067
CAS Number:  
undefined
Protein quantitation reagent, TMTzero™ label reagent
Catalog # PIER90067
CAS Number:  
Specifications
  • Bal.:
    1 KIT
  • Popis:
    TMTzero™ Label Reagent, 5×0,8 mg, sufficient for 5 samples
  • Sekvence:
    01
Specifications
About this item
Pierce™ TMTzero™ Label Reagent serves as a "light" tag for MS-level peptide quantitation similar to duplex isotopic metabolic labeling, such as SILAC, or isotopic dimethylation labeling.