Pierce™ NHS-Fluorescein antibody labeling kit, Pierce™
Katalogové číslo PIER53029
Dodavatel: Thermo Fisher Scientific
Číslo CAS:
Specifikace
- Bal.:1 KIT
- Popis:NHS-Fluorescein antibody labeling kit, sufficient to label and purify 3×1 mg of IgG
Specifikace
O tomto produktu
Pierce™ NHS-Fluorescein antibody labeling kit is a multi-component kit containing a high-performance derivative of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.
- Fluorescently label and purify protein in approximately one hour
- Amine specific labeling - the isothiocyanate variety of fluorescein efficiently labels antibodies and other purified proteins at primary amines
- Single-use vials of reagent for added convenience
Delivery: Standard kit includes NHS Fluorescein (3×50 µg vials), borate buffer (1 ml), spin columns (6 each), collection tubes (12 each) and purification resin (5 ml).