ContactUsLinkComponent
Pycnometer acc. to Hubbard
Pycnometer acc. to Hubbard
Catalog # VTRB632431762023
Supplier:  Kavalier
undefined
Pycnometer acc. to Hubbard
Catalog # VTRB632431762023
Supplier:  Kavalier
Specifications
  • Popis:
    Pycnometer, 25 ml
  • Bal.:
    1
Specifications
About this item
SIMAX® borosilicate glass
For viscous liquids, pastes such as petroleum jelly, creams and bituminous masses.