Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Penicillin/Streptomycin
Penicillin : Streptomycin solution 100X, HyClone™
Penicillin : Streptomycin solution 100X, HyClone™
Penicillin : Streptomycin solution 100X, HyClone™
SV30010
:
HyClone products (Cytiva)
:
Penicillin : Streptomycin solution 100X, HyClone™
SV30010
:
HyClone products (Cytiva)
:
Bal.:
100 ml
Sekvence:
2440
:
:
1406-05-9 (Penicillin) - 57-92-1 (Streptomycin)
:
113-98-4