Reserpine standard for LC-MS, Pierce™
Catalog # PIER88326
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Pierce™ reserpine standard for LC-MS, sufficient for 5 to 500 injections
Specifications
About this item
Reserpine standard for LC-MS is a precise concentration of reserpine specifically designed for performance evaluation of mass spectrometers.
- Pre-diluted and requires minimal additional dilutions to reach target concentration for injection
- Easily removed and replaced with safe handling when withdrawing solution
- Mass spectrometry grade reagent in a non leachable screw cap vials
The standard is provided in a concentration of 100 pg/µl in 50% isopropyl alcohol and requires minimal additional dilutions. The product is provided in a pack of 5×1 ml glass amber vials with PTFE-lined screw caps.