Visual colour kits, chlorine dioxide, MQuant®, Supelco®
1.18754.0001
: Merck
:
- Chemická metoda:DPD
- Bal.:300 Tests
- Popis:Chlorine dioxide test
- M-Clarity quality level:MQ100
- Typ:MCC
- Rozsah:0,020-0,050-0,075-0,10-0,15-0,20-0,30-0,40-0,55 mg/l ClO₂
Quick tests, visual colour comparison.
- Results in approximately 5 minute
- Easy to use
- Application-oriented measuring ranges
- Portable
- A range of economical refill packs (RP) are available
MQuant® with colour card comparator
By using cuvettes with a long light path, the MQuant® (MCC) test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.
: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart or comparator wheel and extensive instructions for use.