Orders LinkContactUsLinkComponent
NMR pipettes, KIMBLE®
NMR pipettes, KIMBLE®
02, NMR pipette, with extended tip (229 mm)
Catalog # SCERSP897085-0009
Supplier:   DWK Life Sciences
NMR pipettes, KIMBLE®
Catalog # SCERSP897085-0009
Supplier:  DWK Life Sciences
NMR pipettes, KIMBLE®
NMR pipettes, KIMBLE®
02, NMR pipette, with extended tip (229 mm)
Přejít na:
Price
5 260,00 Kč
Přihlásit se pro dostupnost
1 * 100 KS
Product Details & Documents
Manufactured from 51 expansion borosilicate glass conforming to USP type I and ASTM E438, type I, class B requirements.
These extended tip NMR pipettes are for retrieving and adding samples in NMR tubes.
Specifikace
  • Popis:
    NMR pipette, with extended tip (229 mm)
  • Sekvence:
    02