Nitrite kits, TNT and TNT plus kit
Catalog # 705-0163
Supplier: Hach
CAS Number:
Specifications
- Chemická metoda:Diazotisation
- Bal.:1 KIT
- Popis:NitriVer 3 TNT Reagent Set
- Sekvence:02
- Pro jednotku:DR 900
- Rozsah:0,003 - 0,500 mg/l
Specifications
About this item
These 'Test N Tube' kits are used for the determination of nitrite by the diazotization method.
- Test 'N Tube products provide safe, convenient testing TNT plus vials reduce errors with bar coded vials and RFID TNT plus vials require no reagent blank